Skip to Content
MilliporeSigma
All Photos(1)

Documents

13005

Millipore

Sulfate API Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Sulfate Reducing API Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

sterility

non-sterile

Quality Level

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 14 g/L
ascorbic acid, 0.1 g/L
dipotassium phosphate, 0.01 g/L
ferrous ammonium sulfate, 0.1 g/L
magnesium sulfate, 0.2 g/L
sodium chloride, 10 g/L
yeast extract, 1 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | metabolite detection: suitable
microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

agriculture
biofuels
environmental
food and beverages
petroleum

microbiology

suitability

Desulfobacter spp.
Desulfovibrio spp.
Desulfuromonas spp.
Thiobacillus spp.
sulfate-reducing bacteria

Application

Sulfate API medium is used for detection and estimation of sulfate reducing bacteria as per American Petroleum Institute Recommended Practice. These bacteria are able to grow and reduce sulfate to sulfide in this medium.

Preparation Note

Suspend 25.4 g in 1000 ml of purified water. Add 4 ml of sodium lactate (Cat. No. 71718; 60% in water). Heat with frequent agitation to dissolve the medium completely. Dispense, preferably in screw-capped tubes in 9 ml amounts. Sterilize at 121°C for 10 minutes. Close the caps immediately while the medium is still hot.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service