Skip to Content
MilliporeSigma
All Photos(1)

Documents

12832

Sigma-Aldrich

3-(2-Benzoxazolyl)umbelliferyl acetate

for fluorescence, ≥98.0% (HPLC)

Synonym(s):

7-Acetoxy-3-(2-benzoxazolyl)coumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H11NO5
CAS Number:
Molecular Weight:
321.28
Beilstein:
5985817
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for fluorescence

Assay

≥98.0% (HPLC)

mp

203-205 °C (lit.)

solubility

DMF: soluble
DMSO: soluble
dioxane: soluble

fluorescence

λex 410 nm; λem 468 nm in 50 mM Tris pH 8.0 (esterase)

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccc2C=C(C(=O)Oc2c1)c3nc4ccccc4o3

Application

The reagent has absorption maxima in the UV range but weak blue fluorescence; however, enzymatic hydrolysis converts it into highly fluorescent anionic molecule.
suitable as fluorogenic substrate for esterases

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service