Skip to Content
MilliporeSigma
All Photos(1)

Documents

12808

Sigma-Aldrich

1-Benzothiophene

purum, ≥95.0% (GC)

Synonym(s):

Thianaphthene, Thionaphthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6S
CAS Number:
Molecular Weight:
134.20
Beilstein:
80580
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥95.0% (GC)

mp

30-33 °C

solubility

organic solvents: soluble

SMILES string

c1ccc2sccc2c1

Application

1-Benzothiophene was used to study the adsorption of 1-benzothiophene in an organic model solution of hydrodesulfurizated gasoline.

Other Notes

This product has been replaced by T27405-ALDRICH | Thianaphthene 95%
Chemistry of 1-benzothiophenes; Selective derivatization and desulfurization to alkylbenzenes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T.G. Back et al.
The Journal of Organic Chemistry, 57, 1986-1986 (1992)
R.M. Scrowston
Advances in Heterocyclic Chemistry, 29, 171-171 (1981)
Mei Xue et al.
Journal of colloid and interface science, 285(2), 487-492 (2005-04-20)
Adsorption of the organic sulfur compounds thiophene (TP) and 1-benzothiophene (1-BTP) in an organic model solution of hydrodesulfurizated gasoline (heptane with 1 wt% toluene and 0.156 mM (5 ppmw as sulfur) TP or 1-BTP) was studied by a batch method
Eva Tyteca et al.
Journal of chromatography. A, 1358, 145-154 (2014-07-21)
Linear gradient programs are very frequently used in reversed phase liquid chromatography to enhance the selectivity compared to isocratic separations. Multi-linear gradient programs on the other hand are only scarcely used, despite their intrinsically larger separation power. Because the gradient-conformity
Hilal E Toraman et al.
Journal of chromatography. A, 1359, 237-246 (2014-07-30)
The detailed compositional characterization of plastic waste pyrolysis oil was performed with comprehensive two-dimensional GC (GC×GC) coupled to four different detectors: a flame ionization detector (FID), a sulfur chemiluminescence detector (SCD), a nitrogen chemiluminescence detector (NCD) and a time of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service