Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12102

Sigma-Aldrich

Chloramine T trihydrate

meets analytical specification of Ph. Eur., BP, 98-103%

Synonym(s):

N-Chloro-p-toluenesulfonamide sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NClNa · 3H2O
CAS Number:
Molecular Weight:
281.69
Beilstein:
3924168
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Quality Level

Assay

98-103%

quality

meets analytical specification of Ph. Eur., BP

reaction suitability

reagent type: oxidant

impurities

ortho compounds, complies
≤2% insoluble in ethanol

pH

8-10 (20 °C, 5%)

mp

167-170 °C (lit.)

suitability

complies for appearance of solution

SMILES string

O.O.O.Cc1ccc(cc1)S(=O)(=O)N([Na])Cl

InChI

1S/C7H7ClNO2S.Na.3H2O/c1-6-2-4-7(5-3-6)12(10,11)9-8;;;;/h2-5H,1H3;;3*1H2/q-1;+1;;;

InChI key

NZYOAGBNMCVQIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

377.6 °F - closed cup

Flash Point(C)

192 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service