Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

108308

Sigma-Aldrich

N-Ethyl-o-toluidine

97%

Synonym(s):

2-(Ethylamino)toluene, N-Ethyl-2-aminotoluene, N-Ethyl-2-methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NHC2H5
CAS Number:
Molecular Weight:
135.21
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Quality Level

Assay

97%

color

colorless

refractive index

n20/D 1.547 (lit.)

bp

218 °C (lit.)

density

0.938 g/mL at 25 °C (lit.)

application(s)

hematology
histology

storage temp.

room temp

SMILES string

CCNc1ccccc1C

InChI

1S/C9H13N/c1-3-10-9-7-5-4-6-8(9)2/h4-7,10H,3H2,1-2H3

InChI key

MWOUGPLLVVEUMM-UHFFFAOYSA-N

Application

N-Ethyl-o-toluidine (2-(Ethylamino)toluene) is a stain used for proteomics research.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ingrid Miller et al.
Proteomics, 6(20), 5385-5408 (2006-09-23)
This review recollects literature data on sensitivity and dynamic range for the most commonly used colorimetric and fluorescent dyes for general protein staining, and summarizes procedures for the most common PTM-specific detection methods. It also compiles some important points to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service