Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09996

Sigma-Aldrich

Zonyl® FS-300 fluorosurfactant

~40% solids in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RfCH2CH2O(CH2CH2O)xH
CAS Number:
MDL number:
UNSPSC Code:
12352300

description

non-ionic

concentration

~40% solids in H2O

density

1.10 g/mL±0.05 g/mL at 20 °C

Legal Information

Zonyl is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service