Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09656

Sigma-Aldrich

1-Hydroxybenzotriazole polymer-bound

100-300 mesh, extent of labeling: ~1.0 mmol/g loading, matrix polystyrene, crosslinked with 2% DVB

Synonym(s):

1-Hydroxybenzotriazole-6-carboxamide bound to polystyrene crosslinked with DVB, HOBt polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: solution phase peptide synthesis

Quality Level

extent of labeling

~1.0 mmol/g loading

matrix

polystyrene, crosslinked with 2% DVB

particle size

100-300 mesh

Other Notes

Polymer bound HOBt for the synthesis of amides and peptides with carbodiimides and other activating reagent without racemization

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Dendrinos et al.
Chemical Communications (Cambridge, England), 499-499 (1998)
Iuliana E. Pop et al.
The Journal of organic chemistry, 62(8), 2594-2603 (1997-04-18)
The synthesis of a new polymer-supported coupling reagent derived from 1-hydroxybenzotriazole is described. An aminomethylated polystyrene was functionalized by reaction with 3-nitro-4-chlorobenzenesulfonyl chloride (2) followed by treatement with hydrazine hydrate, to give the polymeric N-benzyl-1-hydroxybenzotriazole-6-sulfonamide (4).The polymeric reagent 4 was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service