Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09454

Sigma-Aldrich

2-Bromo-6-fluoroaniline

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5BrFN
CAS Number:
Molecular Weight:
190.01
Beilstein:
8541957
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
EC Index Number:
464-930-2
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.582 (lit.)

density

1.674 g/mL at 20 °C (lit.)

functional group

bromo
fluoro

SMILES string

Nc1c(F)cccc1Br

InChI

1S/C6H5BrFN/c7-4-2-1-3-5(8)6(4)9/h1-3H,9H2

InChI key

ALZFPYUPNVLVQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service