Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09222

Sigma-Aldrich

(S)-2-Amino-1-phenylethanol

≥97.0% (sum of enantiomers, GC)

Synonym(s):

(S)-α-(Aminomethyl)benzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO
CAS Number:
Molecular Weight:
137.18
Beilstein:
2412760
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (sum of enantiomers, GC)

optical activity

[α]20/D +43±2°, c = 2% in ethanol

SMILES string

NC[C@@H](O)c1ccccc1

InChI

1S/C8H11NO/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8,10H,6,9H2/t8-/m1/s1

InChI key

ULSIYEODSMZIPX-MRVPVSSYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service