Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

08968

Sigma-Aldrich

Atto 635

BioReagent, suitable for fluorescence, ≥90% (HPCE)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H35ClN2O6
Molecular Weight:
531.04
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioReagent

Assay

≥90% (HPCE)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 635 nm; λem 659 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

[O-]Cl(=O)(=O)=O.CC1=CC(C)(C)N(CCCC(O)=O)c2cc3c(C=C4C=C\C(C=C4C3(C)C)=[N+](/C)C)cc12

InChI

1S/C28H34N2O2.ClHO4/c1-18-17-27(2,3)30(12-8-9-26(31)32)25-16-24-20(14-22(18)25)13-19-10-11-21(29(6)7)15-23(19)28(24,4)5;2-1(3,4)5/h10-11,13-17H,8-9,12H2,1-7H3;(H,2,3,4,5)

InChI key

KIDFITUZQAFBTK-UHFFFAOYSA-N

General description

Atto 635 is a new label with high molecular absorption (120,000) and quantum yield (0.25) as well as sufficient Stokes shift between excitation and emission maximum. This label is optimized for use with diode laser excitation at 635 nm and is characterized by high photostability.

Application

Atto fluorescent labels are designed for high sensitivity applications, including single molecule detection. Atto labels have rigid structures that do not show any cis-trans-isomerization. Thus these labels display exceptional intensity with minimal spectral shift on conjugation.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service