Skip to Content
MilliporeSigma
All Photos(1)

Documents

07958

Supelco

Florisil® suitability check according DIN EN 14039/ISO 16703, Kit

Synonym(s):

Alkane Florisil® suitability check according DIN EN 14039/ISO 16703, Kit, Mineral oil Florisil® suitability check according DIN EN 14039/ISO 16703, Kit, Waste hydrocarbon Florisil® suitability check according DIN EN 14039/ISO 16703, Kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41113319

application(s)

environmental

Components

kit contains:
stearyl stearate, mineral oil standard for calibration 8 mg/ml in heptane

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

24.8 °F

Flash Point(C)

-4.0 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service