Skip to Content
MilliporeSigma
All Photos(1)

Documents

07033

Sigma-Aldrich

N,N-Bis-((S)-1-phenylethyl)dibenzo[d,f][1,3,2]dioxaphosphepin-6-amine

≥99.0%

Synonym(s):

(S,S)-N-(5,7-Diox-6-phosphadibenzo[a,c]cyclohepten-6-yl)bis(1-phenylethyl)amine, O,O′-(2,2′-Biphenyldiyl) N,N-bis[(S)-1-phenylethyl]phosphoramidite, BIPOL-A1(S)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H26NO2P
CAS Number:
Molecular Weight:
439.49
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0%

form

powder

optical activity

[α]/D -250±10°, c = 1 in toluene

reaction suitability

reaction type: click chemistry

mp

99-102 °C

SMILES string

C[C@H](N([C@@H](C)c1ccccc1)P2Oc3ccccc3-c4ccccc4O2)c5ccccc5

InChI

1S/C28H26NO2P/c1-21(23-13-5-3-6-14-23)29(22(2)24-15-7-4-8-16-24)32-30-27-19-11-9-17-25(27)26-18-10-12-20-28(26)31-32/h3-22H,1-2H3

InChI key

JISGHECLGYELKD-UHFFFAOYSA-N

Application

N,N-Bis-((S)-1-phenylethyl)dibenzo[d,f][1,3,2]dioxaphosphepin-6-amine can be used:
  • In the iridium catalyzed allylic arylation.
  • To prepare a copper complex (Cu2X2L3), which is used to detect the transmetalation intermediates in asymmetric addition reactions using ZnEt2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elusive Transmetalation Intermediate in Copper-Catalyzed Conjugate Additions: Direct NMR Detection of an Ethyl Group Attached to a Binuclear Phosphoramidite Copper Complex
von Rekowski F, et al.
Journal of the American Chemical Society, 136(32), 11389-11395 (2014)
Enantioselective Iridium-Catalyzed Allylic Arylation
Polet D, et al.
Chemistry?A European Journal , 15(5), 1205-1216 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service