Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06755

Supelco

Isobutylamine

analytical standard

Synonym(s):

1-Amino-2-methylpropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2NH2
CAS Number:
Molecular Weight:
73.14
Beilstein:
385626
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

refractive index

n20/D 1.396-1.398
n20/D 1.397 (lit.)

bp

64-71 °C (lit.)

mp

−85 °C (lit.)

density

0.736 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(C)CN

InChI

1S/C4H11N/c1-4(2)3-5/h4H,3,5H2,1-2H3

InChI key

KDSNLYIMUZNERS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

15.8 °F - closed cup

Flash Point(C)

-9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirlinda Biba et al.
Journal of chromatography. A, 1363, 250-256 (2014-08-02)
Fast chiral supercritical fluid chromatography (SFC) separations have become important due to the increasing use of high-throughput experimentation (HTE) in organic synthesis. These HTE experiments can generate hundreds of samples for chiral analysis that need to be assayed in a
Lars Erik Uggerhøj et al.
FEBS letters, 588(17), 3291-3297 (2014-07-27)
The inclusion of peptoid monomers into antimicrobial peptides (AMPs) increases their proteolytic resistance, but introduces conformational flexibility (reduced hydrogen bonding ability and cis/trans isomerism). We here use NMR spectroscopy to answer how the insertion of a peptoid monomer influences the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service