Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06060

Sigma-Aldrich

Allyl phenyl sulfide

≥96.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2SC6H5
CAS Number:
Molecular Weight:
150.24
Beilstein:
1856940
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥96.5% (GC)

refractive index

n20/D 1.572

bp

223-225 °C (lit.)

density

1.024 g/mL at 20 °C (lit.)

SMILES string

C=CCSc1ccccc1

InChI

1S/C9H10S/c1-2-8-10-9-6-4-3-5-7-9/h2-7H,1,8H2

InChI key

QGNRLAFFKKBSIM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service