Skip to Content
MilliporeSigma
All Photos(1)

Documents

05983

Sigma-Aldrich

O-Allylhydroxylamine hydrochloride

≥98.0% (AT)

Synonym(s):

O-(2-Propenyl)hydroxylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2ONH2 · HCl
CAS Number:
Molecular Weight:
109.55
Beilstein:
3552394
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (AT)

mp

~170 °C (dec.)

SMILES string

Cl.NOCC=C

InChI

1S/C3H7NO.ClH/c1-2-3-5-4;/h2H,1,3-4H2;1H

InChI key

XIQUJVRFXPBMHS-UHFFFAOYSA-N

Other Notes

Useful intermediate; e.g. cyclization of O-allyloximes to give pyridines

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Koyama et al.
Chemical & Pharmaceutical Bulletin, 31, 2601-2601 (1983)
T. Kusumi et al.
Synthesis, 221-221 (1979)
H. Irie et al.
Heterocycles, 12, 771-771 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service