Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05818

Supelco

4-Allylanisole

analytical standard

Synonym(s):

p-Allylphenyl methyl ether, p-Methoxyallylbenzene, Chavicol methyl ether, Estragole, Isoanethole, Methyl chavicol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2C6H4OCH3
CAS Number:
Molecular Weight:
148.20
Beilstein:
1099454
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.521 (lit.)
n20/D 1.521

bp

215-216 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

COc1ccc(CC=C)cc1

InChI

1S/C10H12O/c1-3-4-9-5-7-10(11-2)8-6-9/h3,5-8H,1,4H2,2H3

InChI key

ZFMSMUAANRJZFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Allylanisole is one of the oxygenated aromatic biogenic volatile organic compounds (BVOCs) which is usually synthesized by plants.

Application

4-Allylanisole has been used as a reference standard for the analysis of 4-allylanisole in essential oils by high performance liquid chromatography (HPLC) equipped with fluorometric detector and in emissions from live vegetation by solid-phase microextraction (SPME) combined with proton transfer reaction mass spectrometry (PTR-MS). It may be used as a reference standard for the determination of 4-allylanisole in food products by simultaneous distillation-extraction (SDE) followed by analyses using capillary gas chromatography (GC)-flame ionization detector (FID) and GC-MS.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of fragrance allergens in essential oils and evaluation of their in vitro permeation from essential oil formulations through cultured skin.
Wang LH and Liu HJ
Journal of Analytical Chemistry, 67(1), 64-71 (2012)
Methyl chavicol: characterization of its biogenic emission rate, abundance, and oxidation products in the atmosphere.
Bouvier-Brown NC, et al.
Atmospheric Chemistry and Physics, 9(6), 2061-2074 (2009)
Determination of estragole, safrole and eugenol methyl ether in food products.
Siano F, et al.
Food Chemistry, 81(3), 469-475 (2003)
Wasma Alhusainy et al.
Toxicological sciences : an official journal of the Society of Toxicology, 129(1), 174-187 (2012-06-01)
The alkenylbenzene estragole is a constituent of several herbs and spices. It induces hepatomas in rodents at high doses following bioactivation by cytochrome P450s and sulfotransferases (SULTs) giving rise to the ultimate carcinogenic metabolite 1'-sulfooxyestragole which forms DNA adducts. Methanolic
Yuji Ishii et al.
Chemical research in toxicology, 24(4), 532-541 (2011-03-10)
Estragole (ES) is a natural constituent of several herbs and spices that acts as a carcinogen in the livers of rodents. Given that the proximal electrophilic form of ES with a reactive carbocation is generated by cytochrome P450 and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service