Skip to Content
MilliporeSigma
All Photos(1)

Documents

05724

Supelco

Polydisperse Particle Standard

Particle Size Standard, PS201, 10 x 0.05g, 3-30 μm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

manufacturer/tradename

Whitehouse Scientific Ltd PS201

particle size

3-30 μm

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

Looking for similar products? Visit Product Comparison Guide

General description

Polydisperse Particle Standard (PS201, size: 3-30 μm) is ideal for identifying particle size distribution (PSD) profile of a particle system.

Application

Used to calibrate particle size analyzers and particle counters.
It may also be used to measure the precision and reproducibility of laser diffractometer particle analyzer.

Features and Benefits

  • suitable for routine instrument calibration checks, testing and corrections
  • available as a neat sample

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Revisiting the chronology and environmental conditions for the accretion of late Pleistocene-early Holocene Pampean loess (Argentina)
Torre G, et al.
Chemosphere, 213, 105-119 (2019)
Arsenic-bearing phases in South Andean volcanic ashes: Implications for As mobility in aquatic environments
Bia G, et al.
Chemical Geology , 393, 26-35 (2015)
Soil variables that determine lead accumulation in Bidens pilosa L. and Tagetes minuta L. growing in polluted soils.
Salazar M J, et al.
Geoderma, 279, 97-108 (2016)
The paleoclimatic message from the polymodal grain-size distribution of late Pleistocene-early Holocene Pampean loess (Argentina)
Torre G, et al.
Chemosphere, 42, 100563-100563 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service