Skip to Content
MilliporeSigma
All Photos(1)

Documents

04954

Sigma-Aldrich

RBS Sericlean

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103206

technique(s)

HPLC: suitable
LC/MS: suitable
ion chromatography: suitable

density

1.16 g/mL at 20 °C

Legal Information

RBS is a trademark of Chemical Products R. Borghgraef S.A.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service