Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04513

Supelco

N-Isobutyrylglycine

analytical standard

Synonym(s):

N-(2-Methyl-1-oxopropyl)glycine, Isobutyryl glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO3
CAS Number:
Molecular Weight:
145.16
Beilstein:
1762789
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

Biochem/physiol Actions

Isobutyrylglycine is an acyl glycine. Acyl glycines are normally minor metabolites of fatty acids. However, the excretion of certain acyl glycines is increased in several inborn errors of metabolism. In certain cases the measurement of these metabolites in body fluids can be used to diagnose disorders associated with mitochondrial fatty acid beta-oxidation. Acyl glycines are produced through the action of glycine N-acyltransferase (EC 2.3.1.13) which is an enzyme that catalyzes the chemical reaction:acyl-CoA + glycine ↔ CoA + N-acylglycine. Isobutyrylglycine is identified in large amount in urine of patients with isobutyryl-CoA dehydrogenase deficiency. Isobutyryl-CoA dehydrogenase deficiency is a disorder caused by the deficiency of isobutyryl-CoA dehydrogenase that is involved in the catabolism of the branched-chain amino acid valine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J O Sass et al.
Journal of inherited metabolic disease, 27(6), 741-745 (2004-10-27)
Isobutyryl-CoA dehydrogenase (IBD) is an enzyme involved in the catabolism of the branched-chain amino acid valine. We report a third and a fourth child with IBD deficiency who were both detected during newborn screening with tandem mass spectrometry and so

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service