Skip to Content
MilliporeSigma
All Photos(1)

Documents

04350

Sigma-Aldrich

3-(N-Ethyl-3-methylanilino)propanesulfonic acid sodium salt

p.a., for enzymic, spectrophotometric determination of H2O2

Synonym(s):

3-(N-Ethyl-m-toluidino)propanesulfonic acid sodium salt, 3-(Ethyl[3-methylphenyl]amino)-1-propanesulfonic acid, N-Ethyl-N-sulfopropyl-m-toluidine sodium salt, Sodium 3-(N-ethyl-3-methylanilino)propanesulfonate, Sodium N-ethyl-N-(3-sulfopropyl)-m-toluidine, TOPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18NNaO3S
CAS Number:
Molecular Weight:
279.33
MDL number:
UNSPSC Code:
12352103

grade

p.a.

quality

for enzymic, spectrophotometric determination of H2O2

technique(s)

UV/Vis spectroscopy: suitable

impurities

5-8% water

SMILES string

[Na+].CCN(CCCS([O-])(=O)=O)c1cccc(C)c1

InChI

1S/C12H19NO3S.Na/c1-3-13(8-5-9-17(14,15)16)12-7-4-6-11(2)10-12;/h4,6-7,10H,3,5,8-9H2,1-2H3,(H,14,15,16);/q;+1/p-1

InChI key

NJZLCEFCAHNYIR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Water-soluble reagent for the enzymatic photometric determination of hydrogen peroxide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamaoku, K., et al.
Analytica Chimica Acta, 136, 121-121 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service