Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04270590

Carvacrol

primary reference standard

Synonym(s):

5-Isopropyl-2-methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H3(CH3)OH
CAS Number:
Molecular Weight:
150.22
Beilstein:
1860514
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

refractive index

n20/D 1.522 (lit.)

bp

236-237 °C (lit.)

mp

3-4 °C (lit.)

density

0.976 g/mL at 20 °C (lit.)

application(s)

food and beverages

storage temp.

−20°C

SMILES string

CC(C)c1ccc(C)c(O)c1

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)10(11)6-9/h4-7,11H,1-3H3

InChI key

RECUKUPTGUEGMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Husnu Can Baser
Current pharmaceutical design, 14(29), 3106-3119 (2008-12-17)
Oregano essential oils obtained from the genera Origanum, Thymus, Coridothymus, Thymbra, Satureja and Lippia are rich in carvacrol, a monoterpenic phenol isomeric with thymol. Turkey is the biggest exporter of oregano herb and oil to the world markets. Oregano is
Antonia Nostro et al.
Recent patents on anti-infective drug discovery, 7(1), 28-35 (2011-11-03)
During the last few years the scientific community has shown a considerable interest in the study of plant materials as sources of new compounds to be processed into antimicrobial agents. In this context, carvacrol, a monoterpenic phenol, has emerged for
C Sánchez et al.
International journal of food microbiology, 192, 72-76 (2014-10-14)
Carvacrol, a monoterpenic phenol, is said to have extensive antimicrobial activity in a wide range of food spoilage or pathogenic fungi, yeast and bacteria. The aim of this study was to assess its antiviral activity on norovirus surrogates, feline calicivirus
Abdollah Khadivi-Khub et al.
Gene, 546(2), 297-308 (2014-06-01)
In this investigation, morphological, phytochemical and ISSR markers were used to estimate the relationships among and within seven populations of white savory (Satureja mutica), belonging to four provinces in Iran. The individuals were phenotypically diverse, which stamen length, corolla length
Laura Higueras et al.
International journal of food microbiology, 188, 53-59 (2014-08-05)
Chitosan/cyclodextrin films (CS:CD) incorporating carvacrol were obtained by casting, and conditioned at 23°C and 75% relative humidity prior to being immersed in liquid carvacrol until they reached sorption equilibrium. In a previous work, the in vitro antimicrobial activity of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service