Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04230

Supelco

Abberior® STAR 470SXP, streptavidin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171501
Pricing and availability is not currently available.

manufacturer/tradename

Abberior GmbH

fluorescence

λex 470 nm; λem 598 nm±10 nm in H2O

suitability

corresponds for SDS-PAGE

detection method

fluorometric

storage temp.

2-8°C

Suitability

Designed and tested for fluorescent super-resolution microscopy

Analysis Note

Unconjugated dye ≤5% of total fluorescence

Legal Information

abberior is a registered trademark of Abberior GmbH

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service