Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03721

Supelco

Nitrite ionophore VI

Selectophore, function tested

Synonym(s):

N,N′-Bis(3,5-di-tert-butylsalicylidene)-1,2-benzenediaminocobalt(II), (SP-4-2)-[[2,2′-[1,2-Phenylenebis[(nitrilo-κN)methylidyne]]bis[4,6-bis(1,1-dimethylethyl)phenolato-κO]](2-)]cobalt, N,N′-Bis-(3,5-di-tert-butylsalicylidene)-o-phenylenediamine cobalt(II), Cobalt(II) tert-butyl-salophen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H46CoN2O2
CAS Number:
Molecular Weight:
597.70
UNSPSC Code:
26111700
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

quality

function tested

λ

in chloroform

UV absorption

λ: 307-317 nm Amax

SMILES string

CC(C)(C)C1=C(O[Co]23O4)C(C=[N]2C5=C([N]3=CC6=C4C(C(C)(C)C)=CC(C(C)(C)C)=C6)C=CC=C5)=CC(C(C)(C)C)=C1

General description

Nitrite ionophore VI based ion selective electrodes (ISEs) are used for the detection of nitrite anions.[1]

Application

Visit our Sensor Applications portal to learn more.
Nitrite ionophore VI has been used in ionophore-based ISE for the detection of nitrite anions by potentiometric sensing.[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitrite-Selective Electrode Based On Cobalt(II) tert-Butyl-Salophen Ionophore.
Neel, B., et al.
Electroanalysis, 26, 473-480 (2014)
Local Acidification of Membrane Surfaces for Potentiometric Sensing of Anions in Environmental Samples.
Pankratova, et al.
ACS sensors, 1(1), 48-54 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service