Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03591

Sigma-Aldrich

Fmoc-D-Arg(Mtr)-OH

≥98.0% (HPLC)

Synonym(s):

Nα-Fmoc-Nω-(4-methoxy-2,3,6-trimethylbenzenesulfonyl)-D-arginine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H36N4O7S
CAS Number:
Molecular Weight:
608.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥98.0% (HPLC)

form

powder

color

white

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

COc1cc(C)c(c(C)c1C)S(=O)(=O)NC(=N)NCCC[C@@H](NC(=O)OCC2c3ccccc3-c4ccccc24)C(O)=O

InChI

1S/C31H36N4O7S/c1-18-16-27(41-4)19(2)20(3)28(18)43(39,40)35-30(32)33-15-9-14-26(29(36)37)34-31(38)42-17-25-23-12-7-5-10-21(23)22-11-6-8-13-24(22)25/h5-8,10-13,16,25-26H,9,14-15,17H2,1-4H3,(H,34,38)(H,36,37)(H3,32,33,35)/t26-/m1/s1

InChI key

LKGHIEITYHYVED-AREMUKBSSA-N

General description

may contain ~5% (w/w) diisopropyl ether

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service