Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03557

Sigma-Aldrich

Poly(ethylene glycol)

tested according to Ph. Eur., 35,000

Synonym(s):

Macrogol 35,000, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

Agency

tested according to Ph. Eur.

Quality Level

form

powder

mol wt

~35000

solubility

aliphatic hydrocarbons: slightly soluble
organic solvents: soluble

application(s)

pharmaceutical (small molecule)

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly (ethylene glycol) can be used as a test compound utilized in the overproduction and purification of recombinant Lactobacillus reuteri LrNDT. It can also be used for concentrating the mutants with lower secretion levels used in ELISAs for the quantitation of shgp190 and of the LIF/shgp190 complexes.

Biochem/physiol Actions

Polyethylene glycol (PEG) is a condensation polymer of ethylene oxide and water. It enhances the refolding of extracted proteins. This enables characterization of crystallized proteins.

Preparation Note

Poly(ethylene glycol) dissolves in many organic solvents and is readily soluble in aromatic hydrocarbons. It also slightly dissolves in aliphatic hydrocarbons.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

281.5 °F - closed cup

Flash Point(C)

138.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jesús Fernández-Lucas et al.
Applied and environmental microbiology, 76(5), 1462-1470 (2010-01-06)
A novel type II nucleoside 2'-deoxyribosyltransferase from Lactobacillus reuteri (LrNDT) has been cloned and overexpressed in Escherichia coli. The recombinant LrNDT has been structural and functionally characterized. Sedimentation equilibrium analysis revealed a homohexameric molecule of 114 kDa. Circular dichroism studies
Juliette Bitard et al.
The Journal of biological chemistry, 278(18), 16253-16261 (2003-02-26)
The leukemia inhibitory factor (LIF) receptor comprises the low affinity binding chain gp190 and the high affinity converter gp130. The ectodomain of gp190 is among the most complex in the hematopoietin receptor family, because it contains two typical cytokine receptor
J L Cleland et al.
The Journal of biological chemistry, 267(19), 13327-13334 (1992-07-05)
Polyethylene glycol (PEG) inhibited aggregation during refolding of bovine carbonic anhydrase B (CAB) through the formation of a nonassociating PEG-intermediate complex. Stoichiometric concentrations of PEG were required for complete recovery of active protein during refolding at aggregating conditions. For example
J L Cleland et al.
Bio/technology (Nature Publishing Company), 10(9), 1013-1019 (1992-09-01)
Previous studies on the refolding of recombinant bovine carbonic anhydrase B (CAB) indicated that polyethylene glycol (PEG) significantly enhanced the recovery of active protein by reducing aggregation. To further test the ability of PEG to enhance refolding, three recombinant human
Idalis Villanueva et al.
Acta biomaterialia, 5(8), 2832-2846 (2009-06-11)
The pericellular matrix (PCM) surrounding chondrocytes is thought to play an important role in transmitting biochemical and biomechanical signals to the cells, which regulates many cellular functions including tissue homeostasis. To better understand chondrocytes interactions with their PCM, three-dimensional poly(ethylene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service