Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03419

Supelco

Piperonal

analytical standard

Synonym(s):

1,3-Benzodioxole-5-carboxaldehyde, 3,4-(Methylenedioxy)benzaldehyde, Heliotropin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6O3
CAS Number:
Molecular Weight:
150.13
Beilstein:
131691
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

1 mmHg ( 87 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

bp

264 °C (lit.)

mp

35-39 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H]C(=O)c1ccc2OCOc2c1

InChI

1S/C8H6O3/c9-4-6-1-2-7-8(3-6)11-5-10-7/h1-4H,5H2

InChI key

SATCULPHIDQDRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Piperonal is a substituted benzaldehyde which is categorized under the class of flavor-related compounds.

Application

Piperonal has been used as a reference standard for the analysis of piperonal in bidi cigarette tobacco by solid phase microextraction-gas chromatography-mass spectrometry (SPME-GC-MS) and in natural vanilla extracts and synthetic flavorings by mixed micellar electrokinetic capillary chromatography (MECC), high performance liquid chromatography (HPLC) and SPME-GC-MS.
It may be used as a reference standard for the determination of piperonal in urban wastewater samples by two-dimensional GC and in commercial coffee granules as aroma compounds by simultaneous multiple SPME-single shot GC-MS.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

150.1 °F

Flash Point(C)

65.62 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of flavour components in natural vanilla extracts and synthetic flavourings by mixed micellar electrokinetic capillary chromatography.
Boyce, et al.
Analytica Chimica Acta, 485(2), 179-186 (2003)
Development of a simultaneous multiple solid-phase microextraction-single shot-gas chromatography/mass spectrometry method and application to aroma profile analysis of commercial coffee.
Lee, et al.
Journal of Chromatography A, 1295(12), 24-41 (2013)
Use of comprehensive two-dimensional gas chromatography for the broad screening of hazardous contaminants in urban wastewaters..
Semard, et al.
Water Science and Technology, 57(12), 179-186 (2008)
S B Stanfill et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(2), 303-317 (2002-12-14)
Indian-made bidi cigarettes sold in the United States are available in a variety of exotic (e.g. clove, mango) and candy-like (e.g. chocolate, raspberry) flavors. Because certain tobacco flavorings contain alkenylbenzenes and other toxic or carcinogenic chemicals, we measured the concentration
Huilin Liu et al.
Journal of agricultural and food chemistry, 63(9), 2545-2549 (2015-02-24)
A novel optosensing chip was constructed by anchoring the fluorescence sensing material layer based on molecularly imprinted polymer (MIP) coated CdSe/ZnS quantum dots (QDs) on a chip for highly selective and sensitive optosensing of sesamol in sesame oil. Many factors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service