Skip to Content
MilliporeSigma
All Photos(1)

Documents

03225

Sigma-Aldrich

1,2,2,6,6-Pentamethyl-4-piperidinol

≥99.0% (GC)

Synonym(s):

4-Hydroxy-1,2,2,6,6-pentamethylpiperidine, HOPEMP, NSC 364075

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H21NO
CAS Number:
Molecular Weight:
171.28
Beilstein:
1498
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (GC)

impurities

≤1% water

mp

72-76 °C

SMILES string

CN1C(C)(C)CC(O)CC1(C)C

InChI

1S/C10H21NO/c1-9(2)6-8(12)7-10(3,4)11(9)5/h8,12H,6-7H2,1-5H3

InChI key

NWHNXXMYEICZAT-UHFFFAOYSA-N

Application

Reactant for:
Synthesis of photostable blue emitting naphthalimides
Synthesis of blue fluorescent naphthalimide pH chemosensors
Synthesis of crowded piperidines with intramolecularly hydrogen-bonded nitrogen
Synthesis of spiropyrans and spirooxazines
Amine photoredox reactions

Other Notes

Building block for introducing a very hindered-amine

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Corr. 1A - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

244.4 °F

Flash Point(C)

118 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R. Mani et al.
J. Macromol. Sci. Chem., A33, 1217-1217 (1996)
S.M. Desai et al.
Macromolecular Symposia, 169, 121-121 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service