Skip to Content
MilliporeSigma
All Photos(1)

Documents

03124-M

Sigma-Aldrich

Potassium iodide

puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

1 mmHg ( 745 °C)

grade

puriss.

Assay

99.0-100.5% (calc. to the dried substance)

form

crystalline

quality

meets analytical specification of Ph. Eur. BP, USP

impurities

ammonium, in accordance
organic volatile impurities, complies (GC)
residual solvents, in accordance
thiosulfate (S2O3), in accordance
≤0.001% heavy metals (as Pb)
≤0.01% thiosulfate (S2O3)
≤0.02% free alkali (as KOH)

loss

≤0.5% loss on drying, 105 °C

pH

6-9 (25 °C, 166 g/L)

mp

681 °C (lit.)

anion traces

iodate (IO3-): in accordance
nitrate (NO3-): in accordance
nitrite (NO2-): in accordance
sulfate (SO42-): ≤0.01%

cation traces

Ba:, in accordance
Fe: ≤0.001%

suitability

complies for appearance of solution

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium iodide (KI) is an inorganic potassium salt. Alumina loaded with potassium iodide has been employed as catalyst for the transesterification of soybean oil with methanol. Influence of KI on the in vitro generation of polymorphonudear leukocyte (PMN)-derived oxygen intermediates has been investigated.

Application

Potassium iodide (KI) was used in the preparation of standard iodide stock solution for the determination of iodide by size exclusion chromatography. It may be used in the spectrophotometric determination of advanced oxidation protein products (AOPP).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Miyachi et al.
The British journal of dermatology, 107(2), 209-214 (1982-08-01)
The effects of potassium iodide, colchicine and dapsone on the in vitro generation of polymorphonuclear leukocyte (PMN)-derived oxygen intermediates were investigated. These three drugs have beneficial effects on those conditions in which PMNs play an important pathogenetic role. Three oxygen
V Witko-Sarsat et al.
Journal of immunology (Baltimore, Md. : 1950), 161(5), 2524-2532 (1998-09-02)
We previously demonstrated the presence of advanced oxidation protein products (AOPP), a novel marker of oxidative stress in the plasma of uremic patients receiving maintenance dialysis. The present study in a cohort of 162 uremic patients showed that plasma concentrations
H B Li et al.
Journal of chromatography. A, 918(2), 335-339 (2001-06-16)
A novel method for the determination of iodide by size exclusion chromatography was established. The method was simple and highly sensitive with good precision. Iodide was converted to iodine, then sequestered with starch, and separated from the matrix using a
Alejandro A Arriagada et al.
Endocrinology, 156(4), 1540-1551 (2015-01-17)
Na+/I- symporter (NIS) mediates iodide (I-) uptake in the thyroid gland, the first and rate-limiting step in the biosynthesis of the thyroid hormones. The expression and function of NIS in thyroid cells is mainly regulated by TSH and by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service