Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

02610

Sigma-Aldrich

Ethyl dimethylphosphonoacetate

technical, ≥90% (GC)

Synonym(s):

Ethoxycarbonylmethylphosphonic acid dimethylester, Phosphonoacetic acid P,P-dimethyl ethylester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2P(O)CH2COOC2H5
CAS Number:
Molecular Weight:
196.14
Beilstein:
1777603
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥90% (GC)

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.435 (lit.)
n20/D 1.435

bp

134 °C/10 mmHg (lit.)

density

1.188 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CP(=O)(OC)OC

InChI

1S/C6H13O5P/c1-4-11-6(7)5-12(8,9-2)10-3/h4-5H2,1-3H3

InChI key

HUNISAHOCCASGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for synthesis of:
  • Non-nucleoside inhibitors of HCV polymerase NS5B
  • Neuropeptide Y1 receptor antagonists
  • Alkenylcarboxylates from Horner-Wadsworth-Emmons reactions

Reactant for:
  • Enantioselective transfer hydrogenation for preparation of the C7-C14 fragment of ulapualide A
  • Intramolecular Michael reactions
  • Olefination of peptidyl aldehydes with stabilized ylides

Other Notes

Horner-Wittig reagent

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hans M Larsson et al.
PloS one, 9(2), e90006-e90006 (2014-03-04)
Although urothelial progenitor-like cells have been described in the human urinary tract, the existence of stem cells remains to be proven. Using a culture system that favors clonogenic epithelial cell growth, we evaluated and characterized clonal human urothelial cells. We
Marie-Anaïs Locquet et al.
Cells, 10(2) (2021-03-07)
Chordomas are rare, slow-growing tumors of the axial skeleton. These tumors are locally aggressive and refractory to conventional therapies. Radical surgery and radiation remain the first-line treatments. Despite these aggressive treatments, chordomas often recur and second-line treatment options are limited.
W.S. Wadsworth
Org. React., 25, 73-73 (1977)
Marjaana Rantala et al.
Bio-protocol, 8(13), e2899-e2899 (2018-07-05)
The hierarchical composition and interactions of the labile thylakoid protein complexes can be assessed by sequential 2D-native gel-electrophoresis system. Mild non-ionic detergent digitonin is used to solubilize labile protein super-and megacomplexes, which are then separated with first-dimension blue native polyacrylamide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service