Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

02379

Supelco

2,3,4,5,6-Pentafluorobenzoic anhydride

for GC derivatization, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14F10O3
CAS Number:
Molecular Weight:
406.13
Beilstein:
3114439
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

Assay

≥98.0% (GC)
≥98.0%

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

mp

66-70 °C

SMILES string

Fc1c(F)c(F)c(c(F)c1F)C(=O)OC(=O)c2c(F)c(F)c(F)c(F)c2F

InChI

1S/C14F10O3/c15-3-1(4(16)8(20)11(23)7(3)19)13(25)27-14(26)2-5(17)9(21)12(24)10(22)6(2)18

InChI key

BGCIWPHADHBSOS-UHFFFAOYSA-N

Other Notes

Derivatizing reagent for lipids

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S T Weintraub et al.
Journal of the American Society for Mass Spectrometry, 11(2), 176-181 (2000-02-26)
Platelet-activating factor is the term used to denote a class of extremely potent lipid mediators that consist predominantly of 1-O-alkyl- and 1-O-acyl-2-acetyl-sn-glycero-3-phosphocholines. A method has been devised for rapid isolation of these acetylated phospholipids by solid-phase extraction prior to direct
D V Crabtree et al.
Journal of chromatography, 466, 251-270 (1989-04-19)
Pentafluorobenzoate derivatives of primary and secondary alcohols have been prepared using pentafluorobenzoic anhydride. The gas chromatographic properties of the homologous series of the methyl esters of 2-hydroxycarboxylic acids from C12 to C26 have been studied on the high-temperature stationary phase
Synthesis of pentafluorobenzoic anhydride: a superior derivatizing agent for lipids.
S T Weintraub et al.
Analytical chemistry, 65(17), 2400-2402 (1993-09-01)
D S Woodard et al.
Biochimica et biophysica acta, 1259(2), 137-147 (1995-11-16)
Platelet-activating factor (PAF), a family of phospholipid autacoids with potent pro-inflammatory activities, is present in saliva. The current study has quantitated various species of PAF isolated from normal human mixed saliva. Choline-containing, sn-2 acetylated phospholipids with sn-1 ether- or ester-linked
B H Chen et al.
Journal of analytical toxicology, 14(1), 12-17 (1990-01-01)
Pentafluoropropionic anhydride (PFPA) and heptafluorobutyric anhydride (HFBA) derivatives of morphine and codeine demonstrated poor spectra due to low abundances of secondary and tertiary ions. Trifluoroacetamide (MBTFA) has been a widely used derivative; however, the internal standard, nalorphine, displayed very poor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service