Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

02374

Supelco

Sodium 1,2-ethanedisulfonate

suitable for ion pair chromatography, ≥99.0% (T)

Synonym(s):

1,2-Ethanedisulfonic acid disodium salt, Disodium 1,2-ethanedisulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaO3SCH2CH2SO3Na
CAS Number:
Molecular Weight:
234.16
Beilstein:
3920126
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NB.21

description

anionic

Quality Level

Assay

≥99.0% (T)

technique(s)

HPLC: suitable
ion pair chromatography: suitable

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: ≤0.1
λ: 220 nm Amax: ≤0.06
λ: 230 nm Amax: ≤0.04
λ: 260 nm Amax: ≤0.02
λ: 500 nm Amax: ≤0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)CCS([O-])(=O)=O

InChI

1S/C2H6O6S2.2Na/c3-9(4,5)1-2-10(6,7)8;;/h1-2H2,(H,3,4,5)(H,6,7,8);;/q;2*+1/p-2

InChI key

BJTHMUJCKBTCFR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Compound for preparation of "edisylate" salts

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service