Skip to Content
MilliporeSigma
All Photos(1)

Documents

02228

Sigma-Aldrich

4-Chloro-1,8-naphthalic anhydride

≥95.0% (T)

Synonym(s):

6-Chloronaphtho[1,8,8a-c,d]pyran-1,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H5ClO3
CAS Number:
Molecular Weight:
232.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥95.0% (T)

mp

202-206 °C (dec.)

storage temp.

2-8°C

SMILES string

Clc1ccc2C(=O)OC(=O)c3cccc1c23

InChI

1S/C12H5ClO3/c13-9-5-4-8-10-6(9)2-1-3-7(10)11(14)16-12(8)15/h1-5H

InChI key

UJEUBSWHCGDJQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefania D'Adamo et al.
Free radical biology & medicine, 153, 159-172 (2020-04-20)
Oxidative stress (OS) contributes to Osteoarthritis (OA) pathogenesis and its effects are worsened by the impairment of homeostatic mechanisms such as autophagy in OA chondrocytes. Rescue of an efficient autophagic flux could therefore reduce the bulk of damaged molecules, and
Cuicui Wang et al.
Bone research, 9(1), 38-38 (2021-08-25)
Glucose metabolism is fundamental for the functions of all tissues, including cartilage. Despite the emerging evidence related to glucose metabolism in the regulation of prenatal cartilage development, little is known about the role of glucose metabolism and its biochemical basis
Cuicui Wang et al.
Bone research, 9(1), 29-29 (2021-06-09)
Atrophic fracture nonunion poses a significant clinical problem with limited therapeutic interventions. In this study, we developed a unique nonunion model with high clinical relevance using serum transfer-induced rheumatoid arthritis (RA). Arthritic mice displayed fracture nonunion with the absence of
Elizabeth A Jolley et al.
Biophysical chemistry, 239, 29-37 (2018-05-29)
DNA duplexes are stabilized by many interactions, one of which is stacking interactions between the nucleic acid bases. These interactions are useful for designing small molecules that bind to DNA. Naphthalimide intercalators have been shown to be valuable anti-cancer agents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service