Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01920

Sigma-Aldrich

Adenosine 5′-(α,β-methylene)diphosphate

≥99.0% (HPLC)

Synonym(s):

α,β-Methyleneadenosine 5′-diphosphate, ADP[α,β-CH2], AMP-CP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17N5O9P2
CAS Number:
Molecular Weight:
425.23
Beilstein:
633678
EC Number:
MDL number:
UNSPSC Code:
41106305

Assay

≥99.0% (HPLC)

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)CP(O)(O)=O)[C@@H](O)[C@H]3O

InChI

1S/C11H17N5O9P2/c12-9-6-10(14-2-13-9)16(3-15-6)11-8(18)7(17)5(25-11)1-24-27(22,23)4-26(19,20)21/h2-3,5,7-8,11,17-18H,1,4H2,(H,22,23)(H2,12,13,14)(H2,19,20,21)/t5-,7-,8-,11-/m1/s1

InChI key

OLCWZBFDIYXLAA-IOSLPCCCSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Powerful inhibitor of 5′-nucleotidase from rat heart membranes; Useful tool in kinetic studies with 5′-nucleotidase

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Naito et al.
The Biochemical journal, 226(3), 645-651 (1985-03-15)
ADP and ATP and their analogues were evaluated as inhibitors of 5'-nucleotidase purified from heart plasma membrane. ADP analogues are more powerful inhibitors than the corresponding ATP analogues. The most powerful inhibitor found is adenosine 5'-[alpha beta-methylene]diphosphate (AOPCP) for which
J. Wierzchowski et al.
Biochimica et Biophysica Acta, 786, 170-170 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service