Skip to Content
MilliporeSigma
All Photos(1)

Documents

01095

Sigma-Aldrich

Bis(trimethylsilyl) acetylenedicarboxylate

≥98.0%

Synonym(s):

Bis(trimethylsilyl) 2-butynedioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O4Si2
CAS Number:
Molecular Weight:
258.42
Beilstein:
4680184
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0%

form

liquid

refractive index

n20/D 1.439 (lit.)
n20/D 1.439

bp

94 °C/0.005 mmHg (lit.)

density

0.999 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)OC(=O)C#CC(=O)O[Si](C)(C)C

InChI

1S/C10H18O4Si2/c1-15(2,3)13-9(11)7-8-10(12)14-16(4,5)6/h1-6H3

InChI key

RYHMNBZWRLSKHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Versatile synthetic intermediate with several advantages (e.g. mild cleavage of the TMS-group)

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K R Jones et al.
Journal of neurocytology, 27(1), 45-57 (1998-04-08)
Alzheimer's disease is thought to be characterized by conformational and phosphorylation changes in tau protein, leading to the formation of aggregations of paired helical filaments within neurons. Potential agents for inducing conformational changes in tau, namely aluminium and glutamate, were
Ilaria Rosso et al.
International journal of molecular sciences, 21(24) (2020-12-30)
Because of their intrinsic characteristics, telomeres are genomic loci that pose significant problems during the replication of the genome. In particular, it has been observed that telomeres that are maintained in cancer cells by the alternative mechanism of the lengthening
Katarzyna J Duda et al.
Nucleic acids research, 49(10), 5568-5587 (2021-05-18)
Heterochromatin has essential functions in maintaining chromosome structure, in protecting genome integrity and in stabilizing gene expression programs. Heterochromatin is often nucleated by underlying DNA repeat sequences, such as major satellite repeats (MSR) and long interspersed nuclear elements (LINE). In
Judit Domingo-Prim et al.
Nature communications, 10(1), 2135-2135 (2019-05-16)
The exosome is a ribonucleolytic complex that plays important roles in RNA metabolism. Here we show that the exosome is necessary for the repair of DNA double-strand breaks (DSBs) in human cells and that RNA clearance is an essential step

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service