Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01032

Sigma-Aldrich

Acetyl coenzyme A trisodium salt

≥90% (HPLC)

Synonym(s):

Acetyl-S-CoA Na3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H35N7Na3O17P3S
CAS Number:
Molecular Weight:
875.52
MDL number:
UNSPSC Code:
41106305

Assay

≥90% (HPLC)

storage temp.

−20°C

Other Notes

Review

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E.R. Stadtman
Reflections on Biochemistry: Proc. Int. Symp. Enzym. Mech. Biosynth. Cell Funct., 161-161 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service