Skip to Content
MilliporeSigma
All Photos(1)

Documents

00879

Sigma-Aldrich

8-Acetoxypyrene-1,3,6-trisulfonic acid trisodium salt

for fluorescence, ≥98.0% (TLC)

Synonym(s):

1-Acetoxypyrene-3,6,8-trisulfonic acid trisodium salt, Trisodium-8-acetoxypyrene-1,3,6-trisulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H9Na3O11S3
CAS Number:
Molecular Weight:
566.42
Beilstein:
9538267
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for fluorescence

Assay

≥98.0% (TLC)

mp

238-248 °C (dec.) (lit.)

solubility

DMF: soluble
DMSO: soluble
H2O: soluble
methanol: soluble

fluorescence

λex 460 nm; λem 510 nm in 0.1 M potassium phosphate pH 8.0 (esterase)

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].CC(=O)Oc1cc(c2ccc3c(cc(c4ccc1c2c34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

Looking for similar products? Visit Product Comparison Guide

Application

A membrane permeable, nonfluorescent reagent . This fluorochrome enters the cell cytoplasm, where its acetic group is cleaved by intracellular, nonspecific esterases, which forms the impermeable, fluorescent form of the molecule. Utilized as a phloem-mobile symplasmic tracer having staining characteristics that differ from the commonly used CFDA (5-(6-)carboxyfluorescein diacetate).
suitable as fluorogenic substrate for esterases

Other Notes

Highly water soluble enzyme substrate for the fluorimetric assay of esterases at longwave excitation and emission wavelengths

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O S Wolfbeis et al.
Analytical biochemistry, 129(2), 365-370 (1983-03-01)
A direct and continuous kinetic method for the fluorimetric assay of various hydrolases by using new, highly water-soluble substrates is described. The latter consist of esters of strongly fluorescent 1-hydroxypyren-3,6,8-trisulfonic acid trisodium salt with acetic, butyric, caprylic, and oleic acid.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service