Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00568

Millipore

Acetone

suitable for HPLC, ≥99.8% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

vapor density

2 (vs air)
2 (vs air)

vapor pressure

184 mmHg ( 20 °C)
184 mmHg

Assay

≥99.8% (GC)

shelf life

limited shelf life, expiry date on the label

expl. lim.

13.2 %
8 % (lit.)

technique(s)

HPLC: suitable

impurities

≤0.2% water

evapn. residue

≤0.001%

refractive index

n20/D 1.359 (lit.)
n20/D 1.359

bp

56 °C/760 mmHg (lit.)
56 °C/760 mmHg (lit.)

mp

-94 °C (lit.)
−94 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

solvent strength ε (Al2O3)

0.56

λ

1 cm path, H2O reference

UV absorption

λ: 330 nm Amax: ≤1.0
λ: 350 nm Amax: ≤0.1
λ: 360 nm Amax: ≤0.01

application(s)

environmental
food and beverages

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Hultberg et al.
PloS one, 6(4), e17665-e17665 (2011-04-13)
For efficient prevention of viral infections and cross protection, simultaneous targeting of multiple viral epitopes is a powerful strategy. Llama heavy chain antibody fragments (VHH) against the trimeric envelope proteins of Respiratory Syncytial Virus (Fusion protein), Rabies virus (Glycoprotein) and
Sinisa Bjelic et al.
Journal of molecular biology, 426(1), 256-271 (2013-10-29)
Designed retroaldolases have utilized a nucleophilic lysine to promote carbon-carbon bond cleavage of β-hydroxy-ketones via a covalent Schiff base intermediate. Previous computational designs have incorporated a water molecule to facilitate formation and breakdown of the carbinolamine intermediate to give the
Oleksiy Krupin et al.
Optics express, 21(1), 698-709 (2013-02-08)
Straight long-range surface plasmon waveguides are demonstrated as biosensors for the detection of cells, proteins and changes in the bulk refractive index of solutions. The sensors consist of 5 μm wide 22 nm thick Au stripes embedded in polymer (CYTOP™)
Juan A López-Ráez et al.
Plant science : an international journal of experimental plant biology, 230, 59-69 (2014-12-07)
Apocarotenoids are a class of compounds that play important roles in nature. In recent years, a prominent role for these compounds in arbuscular mycorrhizal (AM) symbiosis has been shown. They are derived from carotenoids by the action of the carotenoid
Len Verbeke et al.
Hepatology (Baltimore, Md.), 59(6), 2286-2298 (2013-11-22)
The farnesoid X receptor (FXR) is a nuclear bile acid receptor involved in bile acid homeostasis, hepatic and intestinal inflammation, liver fibrosis, and cardiovascular disease. We studied the effect of short-term treatment with obeticholic acid (INT-747), a potent selective FXR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service