Skip to Content
MilliporeSigma
All Photos(1)

Documents

00459

Supelco

Nerolidol

mixture of cis and trans, analytical standard

Synonym(s):

3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol, 3-Hydroxy-3,7,11-trimethyl-1,6,10-dodecatriene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCH2CH2C(CH3)(OH)CH=CH2
CAS Number:
Molecular Weight:
222.37
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

Quality Level

grade

analytical standard

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.3% water

refractive index

n20/D 1.478-1.483
n20/D 1.479 (lit.)

bp

114 °C/1 mmHg (lit.)

density

0.875 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

C\C(C)=C/CC\C(C)=C\CCC(C)(O)C=C

InChI

1S/C15H26O/c1-6-15(5,16)12-8-11-14(4)10-7-9-13(2)3/h6,9,11,16H,1,7-8,10,12H2,2-5H3/b14-11+

InChI key

FQTLCLSUCSAZDY-SDNWHVSQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nerolidol is a naturally occurring sesquiterpene found in the essential oils.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

262.4 °F - closed cup

Flash Point(C)

128 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Lapczynski et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 11, S247-S250 (2008-07-22)
A toxicologic and dermatologic review of nerolidol when used as a fragrance ingredient is presented.
A Lapczynski et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 11, S245-S246 (2008-07-22)
A toxicologic and dermatologic review cis-nerolidol when used as a fragrance ingredient is presented.
D McGinty et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 48 Suppl 3, S43-S45 (2010-02-10)
An addendum to the toxicologic and dermatologic review of Nerolidol (isomer unspecified) when used as a fragrance ingredient is presented.
Tiago Coelho de Assis Lage et al.
Experimental parasitology, 148, 24-29 (2014-12-03)
Baccharis dracunculifolia DC (common name "alecrim-do-campo" in Brazil) is a plant with widespread distribution in South America that is the botanical origin of green propolis. The aim of this study was to evaluate the chemical composition and acaricidal activity of
J A R Curvelo et al.
Journal of medical microbiology, 63(Pt 5), 697-702 (2014-02-14)
Candidiasis is a major opportunistic fungal infection in humans, and its incidence has increased steadily over the last two decades. Candida albicans, the main species of the genus, has a large arsenal of virulence attributes that contribute to successful infections

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service