Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40-2026

Sigma-Aldrich

Phenol red solution

pH 6.8-8.4

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:

form

liquid

availability

available only in Japan

pH

6.8-8.4

SMILES string

O=S1(C2=CC=CC=C2C(C3=CC=C(O)C=C3)(C4=CC=C(O)C=C4)O1)=O

InChI

1S/C19H14O5S/c20-15-9-5-13(6-10-15)19(14-7-11-16(21)12-8-14)17-3-1-2-4-18(17)25(22,23)24-19/h1-12,20-21H

InChI key

BELBBZDIHDAJOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

pH indicator

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisca Alcaraz-Pérez et al.
BMC biotechnology, 8, 81-81 (2008-10-29)
The dual-luciferase assay has been widely used in cell lines to determine rapidly but accurately the activity of a given promoter. Although this strategy has proved very useful, it does not allow the promoter and gene function to be analyzed
Qi Huang et al.
Journal of microbiological methods, 88(2), 271-274 (2011-12-28)
Race 3, biovar 2 strains of Ralstonia solanacearum are quarantined pathogens in Europe and Canada and Select Agent pathogens in the United States. The biovar classification of R. solanacearum strains is based on their biochemical abilities to utilize a carbohydrate
Chromoendoscopy for H. pylori infection--time for a reaction?
José Carlos Marín-Gabriel et al.
Revista espanola de enfermedades digestivas : organo oficial de la Sociedad Espanola de Patologia Digestiva, 104(1), 1-3 (2012-02-04)
Hirotaka Miyamoto et al.
The Journal of pharmacy and pharmacology, 64(6), 848-854 (2012-05-11)
The aim of this study was to determine the factor changing the hepatic disposition of a drug during hypothermia using a rat liver perfusion system. The livers of male Wistar rats were perfused at 37, 32 or 28°C in the
Helle Lysdahl et al.
Stem cell reviews, 9(2), 132-139 (2012-11-09)
The purpose with this study was to investigate the effect of phenol red (PR) on chondrogenic and osteogenic differentiation of human mesenchymal stem cells (hMSCs). hMSCs were differentiated into chondrogenic and osteogenic directions in DMEM with and without PR for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service