Skip to Content
MilliporeSigma
All Photos(1)

Documents

40-1009

Sigma-Aldrich

Alizarin red S solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

form

liquid

availability

available only in Japan

pH

3.7-5.2

Suitability

for titration

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yonghan He et al.
Nature communications, 11(1), 1996-1996 (2020-04-26)
Small molecules that selectively kill senescent cells (SCs), termed senolytics, have the potential to prevent and treat various age-related diseases and extend healthspan. The use of Bcl-xl inhibitors as senolytics is largely limited by their on-target and dose-limiting platelet toxicity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service