Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37-0525

Sigma-Aldrich

Zinc stearate

SAJ first grade, Zn 10.3-10.7 %

Synonym(s):

Stearic acid zinc salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)16COO]2Zn
CAS Number:
Molecular Weight:
632.33
Beilstein:
3919706
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

SAJ first grade

form

solid

autoignition temp.

1454 °F

composition

Zn, 10.3-10.7%

reaction suitability

reagent type: catalyst
core: zinc

availability

available only in Japan

mp

128-130 °C (lit.)

storage temp.

15-25°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)O[Zn]OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/2C18H36O2.Zn/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2*2-17H2,1H3,(H,19,20);/q;;+2/p-2

InChI key

XOOUIPVCVHRTMJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

530.6 °F

Flash Point(C)

277 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service