Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-5140

Sigma-Aldrich

Triton X-100

CP

Synonym(s):

t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS Number:
Beilstein:
2315025
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

grade

CP

description

non-ionic surfactant

form

liquid

mol wt

micellar avg mol wt 80,000
average mol wt 625

availability

available only in Japan

aggregation number

100-155

CMC

0.2-0.9 mM (20-25°C)

transition temp

cloud point 65 °C
pour point ~7 °C

HLB

13.5

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Widely used non-ionic surfactant for recovery of membrane components under mild non-denaturing conditions.

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

539.6 °F

Flash Point(C)

282 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Friederike Pfeiffer et al.
Acta neuropathologica, 122(5), 601-614 (2011-10-11)
In experimental autoimmune encephalomyelitis (EAE), an animal model for multiple sclerosis (MS), loss of the blood-brain barrier (BBB) tight junction (TJ) protein claudin-3 correlates with immune cell infiltration into the CNS and BBB leakiness. Here we show that sealing BBB
Adam A Strömstedt et al.
Antimicrobial agents and chemotherapy, 53(2), 593-602 (2008-11-26)
Methods for increasing the proteolytic stability of EFK17 (EFKRIVQRIKDFLRNLV), a new peptide sequence with antimicrobial properties derived from LL-37, were evaluated. EFK17 was modified by four d-enantiomer or tryptophan (W) substitutions at known protease cleavage sites as well as by
Andri K Riau et al.
Molecular vision, 15, 335-342 (2009-02-19)
Pterygium is an ocular surface disease of unknown etiology associated with epithelial and fibrovascular outgrowth from the conjunctiva onto the cornea. S100 proteins are calcium-activated signaling proteins that interact with other proteins to modulate biological functions such as cell migration
Lionel Rigottier-Gois et al.
PloS one, 6(12), e29023-e29023 (2011-12-24)
Spread of antibiotic resistance among bacteria responsible for nosocomial and community-acquired infections urges for novel therapeutic or prophylactic targets and for innovative pathogen-specific antibacterial compounds. Major challenges are posed by opportunistic pathogens belonging to the low GC% gram-positive bacteria. Among
Manuela Rinaldi et al.
Veterinary research, 42, 61-61 (2011-05-17)
The mucus layer in the gastrointestinal (GI) tract is considered to be the first line of defense to the external environment. Alteration in mucus components has been reported to occur during intestinal nematode infection in ruminants, but the role of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service