Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-4700

Sigma-Aldrich

Trioctylamine

SAJ first grade, ≥99.0%

Synonym(s):

TOA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)7]3N
CAS Number:
Molecular Weight:
353.67
Beilstein:
1210618
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

SAJ first grade

Assay

≥99.0%

availability

available only in Japan

refractive index

n20/D 1.449 (lit.)

bp

164-168 °C/0.7 mmHg (lit.)
365-367 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCN(CCCCCCCC)CCCCCCCC

InChI

1S/C24H51N/c1-4-7-10-13-16-19-22-25(23-20-17-14-11-8-5-2)24-21-18-15-12-9-6-3/h4-24H2,1-3H3

InChI key

XTAZYLNFDRKIHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1 Oral

Target Organs

Heart

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M L Puttemans et al.
Journal - Association of Official Analytical Chemists, 66(4), 1039-1044 (1983-07-01)
Dyes are determined in gelatin-containing sweets. The gelatin must be eliminated first because it interferes with the normal ion-pair extraction of dyes with tri-n-octylamine to chloroform. Techniques such as precipitation of gelatin with organic solvents, and acid and enzymatic digestion
M L Puttemans et al.
Journal - Association of Official Analytical Chemists, 68(1), 80-82 (1985-01-01)
The sorbate content of commercial yogurt samples is determined by reverse phase liquid chromatography following ion-pair extraction with tri-n-octylamine. Mean recoveries (70-88%), precision (1.1-3.3% RSD), and detection limit of the method are presented for sorbic acid, benzoic acid, and saccharin.
M L Puttemans et al.
Journal - Association of Official Analytical Chemists, 68(1), 143-145 (1985-01-01)
Synthetic dyes were extracted from yogurt by different methods, but all methods had in common a liberation of dyes from the food followed by ion-pair formation with tri-n-octylamine. Extraction with pH 5.5 phosphate buffer gave high recoveries for 5 of
Satoshi Tsukahara et al.
Langmuir : the ACS journal of surfaces and colloids, 24(5), 1673-1677 (2008-02-02)
The dynamics of single giant deoxyribonucleic acid (T4GT7DNA, 165 600 base pairs) molecules was examined near and at the toluene/water, toluene-trioctylamine mixtures/water, and trioctylamine/water interfaces by total internal reflection fluorescence microscopy. The results were considerably affected by the trioctylamine content.
Yongshuang Zhu et al.
Dalton transactions (Cambridge, England : 2003), 41(10), 2935-2940 (2012-01-24)
Cobalt phosphate-based organic-inorganic hybrid nanobelts were successfully synthesized in the presence of trioctylamine (TOA) via a solvothermal system. The inorganic cobalt phosphate layers and organic n-octylamine layers in this structure arranged alternately, both of which were parallel to the axial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service