Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-4200

Sigma-Aldrich

Triethylamine

SAJ special grade, ≥98.0%

Synonym(s):

N,N-Diethylethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
Beilstein:
605283
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
grade:
SAJ special grade
bp:
88.8 °C (lit.)
vapor pressure:
51.75 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

SAJ special grade

vapor density

3.5 (vs air)

vapor pressure

51.75 mmHg ( 20 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

593 °F

expl. lim.

8 %

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.401 (lit.)

pH

12.7 (15 °C, 100 g/L)

bp

88.8 °C (lit.)

mp

−115 °C (lit.)

density

0.726 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)CC

InChI

1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

ZMANZCXQSJIPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumanta Kumar Padhi et al.
Inorganic chemistry, 51(15), 8091-8102 (2012-07-26)
Cyclometalated ruthenium complexes having C(^)N and N(^)C type coordinating ligands with NAD(+)/NADH function have been synthesized and characterized by spectroscopic methods. The variation of the coordinating position of σ-donating carbon atom leads to a drastic change in their properties. Both
Kai Morino et al.
Scientific reports, 5, 8953-8953 (2015-05-21)
Well-trained clinicians may be able to provide diagnosis and prognosis from very short biomarker series using information and experience gained from previous patients. Although mathematical methods can potentially help clinicians to predict the progression of diseases, there is no method
Hannah Coyle et al.
The American journal of geriatric psychiatry : official journal of the American Association for Geriatric Psychiatry, 23(4), 335-359 (2014-07-08)
The aim of this study was to assess the efficacy of cognitive training, specifically computerized cognitive training (CCT) and virtual reality cognitive training (VRCT), programs for individuals living with mild cognitive impairment (MCI) or dementia and therefore at high risk
André R Fajardo et al.
Biomacromolecules, 15(7), 2691-2700 (2014-05-27)
Despite a growing interest in amphiphilic polysaccharide-based diblock copolymers as functional polymeric drug delivery nanosystems, biologically relevant sulfated glycosaminoglycan systems were not yet investigated. Here, we report the synthesis and the self-assembly properties in water of chondroitin sulfate-b-poly(lactic acid) (CS-b-PLA(n)).
Jeffrey W Cary et al.
Fungal genetics and biology : FG & B, 81, 88-97 (2015-06-09)
The genome of the filamentous fungus, Aspergillus flavus, has been shown to harbor as many as 56 putative secondary metabolic gene clusters including the one responsible for production of the toxic and carcinogenic, polyketide synthase (PKS)-derived aflatoxins. Except for the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service