Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-2921

Sigma-Aldrich

Titanium(III) chloride solution

JIS special grade, ≥20.0%

Synonym(s):

Titanium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl3
CAS Number:
Molecular Weight:
154.23
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
grade:
JIS special grade
form:
liquid
Pricing and availability is not currently available.

grade

JIS special grade

form

liquid

reaction suitability

reagent type: catalyst
core: titanium

availability

available only in Japan

concentration

≥20.0%

storage temp.

15-25°C

SMILES string

Cl[Ti](Cl)Cl

InChI

1S/3ClH.Ti/h3*1H;/q;;;+3/p-3

InChI key

YONPGGFAJWQGJC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thao Tran P et al.
Chemosphere, 256, 126952-126952 (2020-05-20)
The adjustment of hydraulic retention time (HRT) and the supplement of titanium(III) nitrilotriacetate (Ti(III)-NTA) as a reducing agent were implemented in an anaerobic downflow hanging sponge (DHS) reactor with the aims to (i) improve nitrogen removal performance and to (ii)
Yongfeng Lin et al.
Environmental science & technology, 51(20), 11588-11596 (2017-09-19)
Environmental occurrence and behaviors of 6:2 chlorinated polyfluoroalkyl ether sulfonate (Cl-6:2 PFESA, with trade name F-53B) have been receiving increased attention recently. Nevertheless, its potential fates under diversified conditions remain concealed. In this study, susceptibility of Cl-6:2 PFESA to reductive
Xingxu Lu et al.
ACS applied materials & interfaces, 11(24), 21515-21525 (2019-05-28)
Supported metal catalysts are one of the major classes of heterogeneous catalysts, which demand good stability in both the supports and catalysts. Herein, layered protonated titanate-derived TiO2 (LPT-TiO2) nanowire arrays were synthesized to support platinum catalysts using different loading processes.
Jun-ichi Matsuo et al.
Organic letters, 12(14), 3266-3268 (2010-06-24)
N-p-Toluenesulfonyl (Ts) aldimines reacted with 3-ethoxycyclobutanones by catalysis of titanium(IV) chloride to afford 2,3-di- or 2,3,3-trisubstituted N-Ts-2,3-dihydro-4-pyridones. Synthesis of (+/-)-bremazocine was efficiently accomplished by using this method.
Palaniappan Kulanthaivel et al.
Drug metabolism and disposition: the biological fate of chemicals, 32(9), 966-972 (2004-08-21)
Phase I oxidative metabolism of nitrogen-containing drug molecules to their corresponding N-oxides is a common occurrence. There are instances where liquid chromatography/tandem mass spectometry techniques are inadequate to distinguish this pathway from other oxidation processes, including C-hydroxylations and other heteroatom

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service