Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-5730

Sigma-Aldrich

Sulfanilamide

JIS special grade, ≥99.7%

Synonym(s):

p-Aminobenzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4SO2NH2
CAS Number:
Molecular Weight:
172.20
Beilstein:
511852
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.7%

form

solid

availability

available only in Japan

mp

164-166 °C (lit.)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

Nc1ccc(cc1)S(N)(=O)=O

InChI

1S/C6H8N2O2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H2,8,9,10)

InChI key

FDDDEECHVMSUSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sevgi Kolayli et al.
Journal of enzyme inhibition and medicinal chemistry, 26(6), 895-900 (2011-03-09)
An α-carbonic anhydrase (CA, EC 4.2.1.1) was purified and characterized kinetically from erythrocytes of the sturgeon Acipenser gueldenstaedti, an endangered species. The sturgeon enzyme (AgCA) showed kinetic parameters for the CO(2) hydration reaction comparable with those of the human erythrocytes
Penghua Wang et al.
Water research, 45(16), 5015-5026 (2011-08-02)
A novel carbon-sensitized and nitrogen-doped TiO2 (C/N-TiO2) was synthesized by a facile sol-gel method using titanium butoxide as both titanium precursor and carbon source, and nitric acid as nitrogen source. The calcination temperature had a great effect on the crystal
Ramazan Demirdağ et al.
Bioorganic & medicinal chemistry, 21(6), 1522-1525 (2012-09-15)
Carbonic anhydrase (CA, EC: 4.2.1.1) was purified from sheep kidney by affinity chromatography on a Sepharose 4B-tyrosine-sulfanilamide column. By means of two consecutive procedures, the enzyme (sCA) was purified 227.61-fold with a yield of 60.75%, and a specific activity of
Fei Liu et al.
Organic letters, 12(4), 868-871 (2010-01-19)
The synthesis of benzofused sultams and/or fluorinated sulfonamides, starting from N-allylic sulfonamides, was performed in superacid HF/SbF(5), through superelectrophilic activation. A dramatic effect of superacid composition on the selectivity of the reaction was shown and applied to the synthesis of
Xian-Long Wang et al.
European journal of medicinal chemistry, 45(10), 4631-4639 (2010-08-17)
A series of novel sulfanilamide-derived 1,2,3-triazole compounds were synthesized in excellent yields via 1,3-dipolar cycloaddition and confirmed by MS, IR and NMR spectra as well as elemental analyses. All the compounds were screened in vitro for their antibacterial and antifungal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service