Skip to Content
MilliporeSigma
All Photos(1)

Documents

28-4680

Sigma-Aldrich

Solvent naphtha

CP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

CP

form

liquid

availability

available only in Japan

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Carc. 1B - Muta. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

78.4 °F

Flash Point(C)

25.8 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service