Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-4480

Sigma-Aldrich

Sodium thiocyanate

JIS special grade, ≥99.0%

Synonym(s):

Sodium isothiocyanate, Sodium rhodanate, Sodium rhodanide, Sodium sulfocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaSCN
CAS Number:
Molecular Weight:
81.07
Beilstein:
3594965
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.0%

form

crystalline

availability

available only in Japan

pH

4.8 (20.1 °C, 1070 g/L)

mp

287 °C (dec.) (lit.)

SMILES string

[Na]SC#N

InChI

1S/CHNS.Na/c2-1-3;/h3H;/q;+1/p-1

InChI key

VGTPCRGMBIAPIM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Romero-Steiner et al.
Clinical and diagnostic laboratory immunology, 12(9), 1029-1035 (2005-09-09)
Determination of antibody avidity measurements can be difficult in human serum depending on the population evaluated. We evaluated three approaches for the determination of antibody avidity for immunoglobulin G (IgG). These approaches were (i) elution of bound antibody with increasing
Vincent Ball et al.
Langmuir : the ACS journal of surfaces and colloids, 21(9), 4129-4137 (2005-04-20)
In this study, we investigate the buildup of PEI-(PSS-PAH)(n) polyelectrolyte multilayers at pH 7.4 in the presence of either NaCl or NaSCN as a supporting electrolyte. It appears that in the presence of increasing thiocyanate concentrations (from 0.1 to 0.5
A S Taisova et al.
Biofizika, 49(6), 1069-1074 (2004-12-23)
The properties of the light-harvesting superantenna of the photosynthesizing bacteria from the new family of green filamentous bacteria Oscillochloridaceae were investigated by optical spectroscopy. The antenna of Oscillochloris trichoides consists of peripheral chlorosomal and membrane subantennas. A method of isolation
John W J Hinrichs et al.
American journal of clinical pathology, 143(4), 573-578 (2015-03-18)
To compare next-generation sequencing (NGS) platforms with mutation-specific analysis platforms in a clinical setting, in terms of sensitivity, mutation specificity, costs, capacity, and ease of use. We analyzed 25 formalin-fixed, paraffin-embedded lung cancer samples of different size and tumor percentage
Supramolecular helix-to-helix induction: a 3D anionic framework containing double-helical strands templated by cationic triple-stranded cluster helicates.
Jin-Zhang Hou et al.
Angewandte Chemie (International ed. in English), 47(9), 1711-1714 (2008-01-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service