Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-4040

Sigma-Aldrich

Sodium salicylate

SAJ special grade, ≥99.5%

Synonym(s):

2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt, Sodium 2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COONa
CAS Number:
Molecular Weight:
160.10
Beilstein:
3732792
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥99.5%
grade:
SAJ special grade
form:
crystalline

grade

SAJ special grade

vapor pressure

<0.1 hPa ( 25 °C)

Assay

≥99.5%

form

crystalline

availability

available only in Japan

pH

6.85 (26.3 °C)

mp

>300 °C (lit.)

SMILES string

[Na+].Oc1ccccc1C([O-])=O

InChI

1S/C7H6O3.Na/c8-6-4-2-1-3-5(6)7(9)10;/h1-4,8H,(H,9,10);/q;+1/p-1

InChI key

ABBQHOQBGMUPJH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

208.9 °F - Pensky-Martens closed cup

Flash Point(C)

98.3 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Zhang et al.
Acta biomaterialia, 8(3), 1316-1322 (2011-12-14)
Poly(amidoamine) (PAMAM) dendrons were prepared from hollow fiber membranes (HFM) consisting of bromomethylated poly(2,6-dimethyl-1,4-phenylene oxide) (BPPO) in a stepwise manner. The prepared HFM were characterized by Fourier transform infrared spectroscopy, elemental analysis, and scanning electron microscopy. The drug loading efficiency
Itsuro Kazama et al.
Immunopharmacology and immunotoxicology, 34(5), 874-878 (2012-03-14)
Lymphocytes predominantly express delayed rectifier K(+)-channels (Kv1.3) in their plasma membranes, and the channels play crucial roles in the lymphocyte activation and proliferation. Since nonsteroidal anti-inflammatory drugs (NSAIDs), the most commonly used analgesic and antipyretic drugs, exert immunomodulatory effects, they
Yan-Yan Su et al.
PloS one, 7(10), e46969-e46969 (2012-10-17)
Sodium salicylate (NaSal), an aspirin metabolite, can cause tinnitus in animals and human subjects. To explore neural mechanisms underlying salicylate-induced tinnitus, we examined effects of NaSal on neural activities of the medial geniculate body (MGB), an auditory thalamic nucleus that
Guang-Di Chen et al.
Hearing research, 295, 100-113 (2012-12-04)
High doses of sodium salicylate (SS) have long been known to induce temporary hearing loss and tinnitus, effects attributed to cochlear dysfunction. However, our recent publications reviewed here show that SS can induce profound, permanent, and unexpected changes in the
Astam K Patra et al.
Journal of hazardous materials, 201-202, 170-177 (2011-12-16)
We report a highly efficient synthetic strategy for self-assembled mesoporous γ-Al(2)O(3) materials using sodium salicylate as template. The mesoporous γ-Al(2)O(3) samples synthesized following this strategy have high surface areas (231-497 m(2)g(-1)), consist of crystalline tiny spherical nanoparticles of dimensions ca.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service