Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-3470

Sigma-Aldrich

Sodium nitrite

SAJ first grade, ≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaNO2
CAS Number:
Molecular Weight:
69.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥97.0%
grade:
SAJ first grade
form:
solid

grade

SAJ first grade

vapor pressure

<0.0000 hPa ( 25 °C)

Assay

≥97.0%

form

solid

autoignition temp.

914 °F

availability

available only in Japan

pH

9

mp

271 °C (lit.)

SMILES string

[Na+].[O-]N=O

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

LPXPTNMVRIOKMN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter J Rix et al.
Clinical pharmacokinetics, 54(3), 261-272 (2014-11-26)
The efficacy of nebulized sodium nitrite (AIR001) has been demonstrated in animal models of pulmonary arterial hypertension (PAH), but it was not known if inhaled nitrite would be well tolerated in human subjects at exposure levels associated with efficacy in
Rustem F Dautov et al.
Nitric oxide : biology and chemistry, 40, 36-44 (2014-05-27)
Previous studies in non-human blood vessels and in platelets have demonstrated that under hypoxic conditions release of NO from nitrite (NO2(-)) is potentiated by deoxyhaemoglobin. In the current study, we characterized hypoxic potentiation of NO2(-) effects in human vasculature and
Edward H Oldfield et al.
Journal of neurosurgery, 119(3), 634-641 (2013-05-28)
Intravenous sodium nitrite has been shown to prevent and reverse cerebral vasospasm in a primate model of subarachnoid hemorrhage (SAH). The present Phase IIA dose-escalation study of sodium nitrite was conducted to determine the compound's safety in humans with aneurysmal
Lucas C Pinheiro et al.
Free radical biology & medicine, 53(4), 701-709 (2012-06-23)
The new pathway nitrate-nitrite-nitric oxide (NO) has emerged as a physiological alternative to the classical enzymatic pathway for NO formation from l-arginine. Nitrate is converted to nitrite by commensal bacteria in the oral cavity and the nitrite formed is then
Borja Ibáñez et al.
Journal of the American College of Cardiology, 65(14), 1454-1471 (2015-04-11)
The damage inflicted on the myocardium during acute myocardial infarction is the result of 2 processes: ischemia and subsequent reperfusion (ischemia/reperfusion injury). During the last 3 decades, therapies to reduce ischemic injury (mainly reperfusion strategies) have been widely incorporated into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service